2-Cyan-3-pyridinboronsäurepinakolester

Lieferant: Apollo Scientific

Synonyme: 2-Cyanopyridine-3-boronic acid, pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile

OR310242-1G
APOSOR310242-1GEA 92.8 EUR
APOSOR310242-1G
2-Cyan-3-pyridinboronsäurepinakolester
2-Cyan-3-pyridinboronsäurepinakolester
Formel: C₁₂H₁₅BN₂O₂
Molecular Weight: 230,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08458477
CAS-Nummer: 878194-93-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR