3-Bromo-2-methoxy-5-methyl-1,6-naphthyridine 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR310250-500MG
APOSOR310250-500MGEA
782
EUR
APOSOR310250-500MG
3-Bromo-2-methoxy-5-methyl-1,6-naphthyridine 97%
3-Bromo-2-methoxy-5-methyl-1,6-naphthyridine
Formel:
C₁₀H₉BrN₂O Schmelzpunkt: 126…128 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD22375452 CAS-Nummer: 1383468-70-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...