1-Benzyl-1,9-diazaspiro[5.5]undecane

Lieferant: Apollo Scientific

OR310253-500MG
APOSOR310253-500MGEA 883 EUR
APOSOR310253-500MG
1-Benzyl-1,9-diazaspiro[5.5]undecane
1-Benzyl-1,9-diazaspiro[5.5]undecane
Formel: C₁₆H₂₄N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12406522
CAS-Nummer: 1100748-68-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR