6-Phenylpyridine-3-boronic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Borono-2-phenylpyridine, (6-Phenylpyridin-3-yl)boronic acid
OR310262-500MG
APOSOR310262-500MGEA
438
EUR
APOSOR310262-500MG
6-Phenylpyridine-3-boronic acid
6-Phenylpyridine-3-boronic acid
Formel:
C₁₁H₁₀BNO₂ Molecular Weight: 199,014 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD03788243 CAS-Nummer: 155079-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...