(1R,2S,5S)-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid ≥99%, ee 99%

Lieferant: Apollo Scientific
Achtung

OR310264-500MG
APOSOR310264-500MGEA 1350 EUR
APOSOR310264-500MG
(1R,2S,5S)-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid ≥99%, ee 99%
(1R,2S,5S)-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid
Formel: C₆H₉NO₂
Schmelzpunkt: 252…256 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05663832
CAS-Nummer: 33294-81-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR