1-Methylnaphthalene-2-boronic acid 97%

Lieferant: Apollo Scientific

Synonyme: (1-Methylnaphthalen-2-yl)boronic acid

OR310268-1G
APOSOR310268-1GEA 319 EUR
APOSOR310268-1G
1-Methylnaphthalene-2-boronic acid 97%
1-Methylnaphthalene-2-boronic acid
Formel: C₁₁H₁₁BO₂
Molecular Weight: 186.015 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10566508
CAS-Nummer: 590401-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR