1-Butyn-3-yl-p-toluenesulphonate
Lieferant: Apollo Scientific
Synonyme:
p-Toluenesulphonic acid 1-Butyn-3-yl Ester, (RS)-1-Methyl-2-propynyl-p-toluenesulphonate, But-3-yn-2-yl 4-methylbenzenesulphonate
OR310300-5G
APOSOR310300-5GEA
190
EUR
APOSOR310300-5G
1-Butyn-3-yl-p-toluenesulphonate
1-Butyn-3-yl-p-toluenesulphonate
Formel:
C₁₁H₁₂O₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060095 CAS-Nummer: 53487-52-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...