4-(Chloromethyl)-1,2-dinitrobenzene
Lieferant: Apollo Scientific
OR310304-5G
APOSOR310304-5GEA
1930
EUR
APOSOR310304-5G
4-(Chloromethyl)-1,2-dinitrobenzene
4-(Chloromethyl)-1,2-dinitrobenzene
Formel:
C₇H₅ClN₂O₄ Molecular Weight: 216,579 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13689020 CAS-Nummer: 89692-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...