(3,4-Diaminophenyl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
OR310306-500MG
APOSOR310306-500MGEA
438
EUR
APOSOR310306-500MG
(3,4-Diaminophenyl)methanol
(3,4-Diaminophenyl)methanol
Formel:
C₇H₁₀N₂O Schmelzpunkt: 97…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173951 CAS-Nummer: 63189-98-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...