4-(Hydroxymethyl)-4-phenylcyclohexan-1-one

Lieferant: Apollo Scientific

OR310320-1G
APOSOR310320-1GEA 538 EUR
APOSOR310320-1G
4-(Hydroxymethyl)-4-phenylcyclohexan-1-one
4-(Hydroxymethyl)-4-phenylcyclohexan-1-one
Formel: C₁₃H₁₆O₂
Molecular Weight: 204,265 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17013064
CAS-Nummer: 51510-01-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR