Methanesulphonic acid amino 4-methoxybenzoate
Lieferant: Apollo Scientific
OR310324-1G
APOSOR310324-1GEA
538
EUR
APOSOR310324-1G
Methanesulphonic acid amino 4-methoxybenzoate
Methanesulphonic acid amino 4-methoxybenzoate
Formel:
C₈H₉NO₃·CH₃SO₃H Molecular Weight: 263,268 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11111599 CAS-Nummer: 872851-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...