2-[4-(Octyloxy)phenyl]thiophene

Lieferant: Apollo Scientific

OR310325-1G
APOSOR310325-1GEA 538 EUR
APOSOR310325-1G
2-[4-(Octyloxy)phenyl]thiophene
2-[4-(Octyloxy)phenyl]thiophene
Formel: C₁₈H₂₄OS
Molecular Weight: 288,448 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26523307
CAS-Nummer: 850893-32-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR