(2S)-2,3-Dimethylbutane-1,3-diol
Lieferant: Apollo Scientific
OR310330-500MG
APOSOR310330-500MGEA
661
EUR
APOSOR310330-500MG
(2S)-2,3-Dimethylbutane-1,3-diol
(2S)-2,3-Dimethylbutane-1,3-diol
Formel:
C₆H₁₄O₂ Molecular Weight: 118,174 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10574870 CAS-Nummer: 73295-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...