5-Bromo-4-chloro-7-(4-methylbenzenesulphonyl)-7H-pyrrolo[2,3-d]pyrimidine

Lieferant: Apollo Scientific

OR310345-1G
APOSOR310345-1GEA 270 EUR
APOSOR310345-1G
5-Bromo-4-chloro-7-(4-methylbenzenesulphonyl)-7H-pyrrolo[2,3-d]pyrimidine
5-Bromo-4-chloro-7-(4-methylbenzenesulphonyl)-7H-pyrrolo[2,3-d]pyrimidine
Formel: C₁₃H₉BrClN₃O₂S
Molecular Weight: 386,651 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20441317
CAS-Nummer: 1143534-78-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR