Ethyl 7-nitro-2,3-dihydro-1-benzofuran-2-carboxylate

Lieferant: Apollo Scientific

OR310347-500MG
APOSOR310347-500MGEA 661 EUR
APOSOR310347-500MG
Ethyl 7-nitro-2,3-dihydro-1-benzofuran-2-carboxylate
Ethyl 7-nitro-2,3-dihydro-1-benzofuran-2-carboxylate
Formel: C₁₁H₁₁NO₅
Molecular Weight: 237,209 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26523315
CAS-Nummer: 1565845-67-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR