N-(4-Methoxy-2-methylquinolin-6-yl)acetamide

Lieferant: Apollo Scientific

OR310348-1G
APOSOR310348-1GEA 538 EUR
APOSOR310348-1G
N-(4-Methoxy-2-methylquinolin-6-yl)acetamide
N-(4-Methoxy-2-methylquinolin-6-yl)acetamide
Formel: C₁₃H₁₄N₂O₂
Molecular Weight: 230,262 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06681161
CAS-Nummer: 100795-23-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR