Benzyl N-[2-(4-hydroxyphenyl)ethyl]carbamate

Lieferant: Apollo Scientific

OR310350-1G
APOSOR310350-1GEA 180 EUR
APOSOR310350-1G
Benzyl N-[2-(4-hydroxyphenyl)ethyl]carbamate
Benzyl N-[2-(4-hydroxyphenyl)ethyl]carbamate
Formel: C₁₆H₁₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD25458437
CAS-Nummer: 29655-46-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR