4'-Amino-[1,1'-biphenyl]-4-carboxylic acid 95+%

Lieferant: Apollo Scientific
Achtung

OR310360-1G
APOSOR310360-1GEA 81.2 EUR
APOSOR310360-1G
4'-Amino-[1,1'-biphenyl]-4-carboxylic acid 95+%
4'-Amino-[1,1'-biphenyl]-4-carboxylic acid
Formel: C₁₃H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01138446
CAS-Nummer: 5730-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR