4'-Formyl-2'-nitroacetanilide

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(4-Formyl-2-nitrophenyl)acetamide, 4-Acetamido-3-nitrobenzaldehyde

OR310363-1G
APOSOR310363-1GEA 130 EUR
APOSOR310363-1G
4'-Formyl-2'-nitroacetanilide
4'-Formyl-2'-nitroacetanilide
Formel: C₉H₈N₂O₄
Molecular Weight: 208.171 g/mol
Schmelzpunkt: 155…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00092015
CAS-Nummer: 51818-98-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR