Ethyl 2-(1,3-dithietan-2-ylidene)-3-oxobutanoate

Lieferant: Apollo Scientific

OR310380-1G
APOSOR310380-1GEA 538 EUR
APOSOR310380-1G
Ethyl 2-(1,3-dithietan-2-ylidene)-3-oxobutanoate
Ethyl 2-(1,3-dithietan-2-ylidene)-3-oxobutanoate
Formel: C₈H₁₀O₃S₂
Molecular Weight: 218,293 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18800140
CAS-Nummer: 120596-96-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR