(2S)-2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}non-8-enoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)non-8-enoic acid, Fmoc-2-(6'-Octenyl)-Gly-OH

OR310381-1G
APOSOR310381-1GEA 336 EUR
APOSOR310381-1G
(2S)-2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}non-8-enoic acid
(2S)-2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}non-8-enoic acid
Formel: C₂₄H₂₇NO₄
Molecular Weight: 393,475 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094931
CAS-Nummer: 1058705-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR