4-(Methylsulfonyl)phenylboronsäurepinakolester

Lieferant: Apollo Scientific

Synonyme: 4,4,5,5-Tetramethyl-2-(4-(methylsulphonyl)phenyl)-1,3,2-dioxaborolane, 2-(4-Methanesulphonylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(Methanesulphonyl)phenylboronic acid, pinacol ester

OR310395-5G
APOSOR310395-5GEA 66.7 EUR
APOSOR310395-5G
4-(Methylsulfonyl)phenylboronsäurepinakolester
4-(Methylsulfonyl)phenylboronsäurepinakolester
Formel: C₁₃H₁₉BO₄S
Molecular Weight: 282,1636 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863921
CAS-Nummer: 603143-27-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR