tert-Butyl 5-(hydroxymethyl)-2,3-dihydro-1H-isoindole-2-carboxylate

Lieferant: Apollo Scientific

OR310406-1G
APOSOR310406-1GEA 671 EUR
APOSOR310406-1G
tert-Butyl 5-(hydroxymethyl)-2,3-dihydro-1H-isoindole-2-carboxylate
tert-Butyl 5-(hydroxymethyl)-2,3-dihydro-1H-isoindole-2-carboxylate
Formel: C₁₄H₁₉NO₃
Molecular Weight: 249,306 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16090011
CAS-Nummer: 253801-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR