Methyl 2-(2-cyano-3-ethoxy-3-oxopropyl)benzoate

Lieferant: Apollo Scientific

OR310407-1G
APOSOR310407-1GEA 538 EUR
APOSOR310407-1G
Methyl 2-(2-cyano-3-ethoxy-3-oxopropyl)benzoate
Methyl 2-(2-cyano-3-ethoxy-3-oxopropyl)benzoate
Formel: C₁₄H₁₅NO₄
Molecular Weight: 261,273 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06211147
CAS-Nummer: 153802-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR