3-Bromthieno[3,2-b]thiophen

Lieferant: Apollo Scientific
Achtung

OR310409-1G
APOSOR310409-1GEA 221 EUR
APOSOR310409-1G
3-Bromthieno[3,2-b]thiophen
3-Bromthieno[3,2-b]thiophen
Formel: C₆H₃BrS₂
Molecular Weight: 219,11 g/mol
Dichte: 1,78 g/cm³ (25 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD10000952
CAS-Nummer: 25121-83-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR