2-Ethyl 5-(4-nitrophenyl)methyl 4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine-2,5-dicarboxylate

Lieferant: Apollo Scientific

Synonyme: 2-Ethyl 5-(4-nitrobenzyl) 6,7-dihydropyrazolo[1,5-a]pyrazine-2,5(4H)-dicarboxylate

OR310410-500MG
APOSOR310410-500MGEA 661 EUR
APOSOR310410-500MG
2-Ethyl 5-(4-nitrophenyl)methyl 4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine-2,5-dicarboxylate
2-Ethyl 5-(4-nitrophenyl)methyl 4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine-2,5-dicarboxylate
Formel: C₁₇H₁₈N₄O₆
Molecular Weight: 374,348 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27924230
CAS-Nummer: 623565-14-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR