1,1-Diethylpropargylamine

Lieferant: Apollo Scientific
Gefahr

OR310413-1G
APOSOR310413-1GEA 336 EUR
APOSOR310413-1G
1,1-Diethylpropargylamine
1,1-Diethylpropargylamine
Formel: C₇H₁₃N
Molecular Weight: 111.185 g/mol
Dichte: 0.824 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008057
CAS-Nummer: 3234-64-8
UN: 2733
ADR: 3+8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR