N-Phenylquinolin-4-amine

Lieferant: Apollo Scientific

OR310414-1G
APOSOR310414-1GEA 347 EUR
APOSOR310414-1G
N-Phenylquinolin-4-amine
N-Phenylquinolin-4-amine
Formel: C₁₅H₁₂N₂
Molecular Weight: 220.269 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00958207
CAS-Nummer: 30696-07-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR