Propan-2-yl 5-bromothiophene-2-carboxylate
Lieferant: Apollo Scientific
OR310417-1G
APOSOR310417-1GEA
671
EUR
APOSOR310417-1G
Propan-2-yl 5-bromothiophene-2-carboxylate
Propan-2-yl 5-bromothiophene-2-carboxylate
Formel:
C₈H₉BrO₂S Molecular Weight: 249,125 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21101334 CAS-Nummer: 1174013-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...