Imidazo[1,2-a]pyridin-2(3H)-one hydrobromide

Lieferant: Apollo Scientific

OR310419-1G
APOSOR310419-1GEA 538 EUR
APOSOR310419-1G
Imidazo[1,2-a]pyridin-2(3H)-one hydrobromide
Imidazo[1,2-a]pyridin-2(3H)-one hydrobromide
Formel: C₇H₆N₂O·BrH
Molecular Weight: 215,047 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00226053
CAS-Nummer: 107934-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR