N-tert-Butyl-5-propylthiophene-2-carboxamide
Lieferant: Apollo Scientific
OR310422-1G
APOSOR310422-1GEA
538
EUR
APOSOR310422-1G
N-tert-Butyl-5-propylthiophene-2-carboxamide
N-tert-Butyl-5-propylthiophene-2-carboxamide
Formel:
C₁₂H₁₉NOS Molecular Weight: 225,35 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27924231 CAS-Nummer: 166591-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...