8-[(4-Methylbenzene)sulphonyl]-1,4-dioxa-8-azaspiro[4.5]decane

Lieferant: Apollo Scientific

OR310426-1G
APOSOR310426-1GEA 336 EUR
APOSOR310426-1G
8-[(4-Methylbenzene)sulphonyl]-1,4-dioxa-8-azaspiro[4.5]decane
8-[(4-Methylbenzene)sulphonyl]-1,4-dioxa-8-azaspiro[4.5]decane
Formel: C₁₄H₁₉NO₄S
Molecular Weight: 297,37 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01252045
CAS-Nummer: 853751-19-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR