1-[4-(2-Chloroethoxy)phenyl]-2-phenylbutan-1-one
Lieferant: Apollo Scientific
OR310428-500MG
APOSOR310428-500MGEA
782
EUR
APOSOR310428-500MG
1-[4-(2-Chloroethoxy)phenyl]-2-phenylbutan-1-one
1-[4-(2-Chloroethoxy)phenyl]-2-phenylbutan-1-one
Formel:
C₁₈H₁₉ClO₂ Molecular Weight: 302,795 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09835550 CAS-Nummer: 103628-22-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...