[4-(Benzyloxy)phenyl]acetylene

Lieferant: Apollo Scientific
Achtung

Synonyme: 4'-Benzyloxyphenyl acetylene

OR31043-1G
APOSOR31043-1GEA 69.6 EUR
APOSOR31043-1G
[4-(Benzyloxy)phenyl]acetylene
[4-(Benzyloxy)phenyl]acetylene
Formel: C₁₅H₁₂O
Molecular Weight: 208,255 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01814885
CAS-Nummer: 84284-70-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR