1-Boc-indoline-5-boronic acid pinacol ester
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-indole-1-carboxylate, tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indoline-1-carboxylate
OR310450-1G
APOSOR310450-1GEA
85.7
EUR
APOSOR310450-1G
1-Boc-indoline-5-boronic acid pinacol ester
1-Boc-indoline-5-boronic acid pinacol ester
Formel:
C₁₉H₂₈BNO₄ Schmelzpunkt: 120…121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12408237 CAS-Nummer: 837392-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...