2-[4-(3-Amino-4-nitrophenyl)piperazin-1-yl]ethan-1-ol

Lieferant: Apollo Scientific

OR310457-5G OR310457-1G
APOSOR310457-5GEA 0 EUR
APOSOR310457-5G APOSOR310457-1G
2-[4-(3-Amino-4-nitrophenyl)piperazin-1-yl]ethan-1-ol
2-[4-(3-Amino-4-nitrophenyl)piperazin-1-yl]ethan-1-ol
Formel: C₁₂H₁₈N₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00545981
CAS-Nummer: 23470-44-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR