Methyl 6-(tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene-2-carboxylate

Lieferant: Apollo Scientific

Synonyme: Methyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-naphthoate

OR310471-1G
APOSOR310471-1GEA 155 EUR
APOSOR310471-1G
Methyl 6-(tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene-2-carboxylate
Methyl 6-(tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene-2-carboxylate
Formel: C₁₈H₂₁BO₄
Molecular Weight: 312,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18783177
CAS-Nummer: 736989-93-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR