5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-dihydro-2-benzofuran-1-one

Lieferant: Apollo Scientific

OR310472-1G
APOSOR310472-1GEA 82.8 EUR
APOSOR310472-1G
5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-dihydro-2-benzofuran-1-one
5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-dihydro-2-benzofuran-1-one
Formel: C₁₄H₁₇BO₄
Molecular Weight: 260,093 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18251723
CAS-Nummer: 862081-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR