3-Bromophenylboronic acid MIDA ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(3-Bromophenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

OR310497-5G
APOSOR310497-5GEA 201 EUR
APOSOR310497-5G
3-Bromophenylboronic acid MIDA ester
3-Bromophenylboronic acid MIDA ester
Formel: C₁₁H₁₁BBrNO₄
Molecular Weight: 311.92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11215220
CAS-Nummer: 1287221-37-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR