Methyl 1-[2-(2H-1,3-benzodioxol-5-yl)ethyl]-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
Lieferant: Apollo Scientific
OR310504-1G
APOSOR310504-1GEA
671
EUR
APOSOR310504-1G
Methyl 1-[2-(2H-1,3-benzodioxol-5-yl)ethyl]-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
Methyl 1-[2-(2H-1,3-benzodioxol-5-yl)ethyl]-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
Formel:
C₁₅H₁₄N₂O₆ Molecular Weight: 318,281 g/mol Schmelzpunkt: 226…228 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD27957327 CAS-Nummer: 1565827-86-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...