S,S-Tetramethylenesulphoximide

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Imino-1-oxotetrahydrothiophene, Tetrahydro-1H-thiophen-1-imine 1-oxide, 1-Iminotetrahydrothiophene 1-oxide

OR310524-1G
APOSOR310524-1GEA 538 EUR
APOSOR310524-1G
S,S-Tetramethylenesulphoximide
S,S-Tetramethylenesulphoximide
Formel: C₄H₉NOS
Molecular Weight: 119,185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27924567
CAS-Nummer: 50578-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR