Methyl 5'-hydroxy-5,5-dimethyl-3',3'a,4',6'a-tetrahydro-1'H-spiro[1,3-dioxane-2,2'-pentalene]-6'-carboxylate

Lieferant: Apollo Scientific

OR310526-500MG
APOSOR310526-500MGEA 560 EUR
APOSOR310526-500MG
Methyl 5'-hydroxy-5,5-dimethyl-3',3'a,4',6'a-tetrahydro-1'H-spiro[1,3-dioxane-2,2'-pentalene]-6'-carboxylate
Methyl 5'-hydroxy-5,5-dimethyl-3',3'a,4',6'a-tetrahydro-1'H-spiro[1,3-dioxane-2,2'-pentalene]-6'-carboxylate
Formel: C₁₅H₂₂O₅
Molecular Weight: 282.332 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27957337
CAS-Nummer: 105448-66-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR