Methyl 5-(4-chloro-3-methylphenyl)-1H-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR310528-1G
APOSOR310528-1GEA 538 EUR
APOSOR310528-1G
Methyl 5-(4-chloro-3-methylphenyl)-1H-pyrazole-3-carboxylate
Methyl 5-(4-chloro-3-methylphenyl)-1H-pyrazole-3-carboxylate
Formel: C₁₂H₁₁ClN₂O₂
Molecular Weight: 250,681 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26387491
CAS-Nummer: 192702-05-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR