Methyl 6-bromo-1-(propan-2-yl)-1H-indazole-4-carboxylate
Lieferant: Apollo Scientific
OR310529-1G
APOSOR310529-1GEA
317
EUR
APOSOR310529-1G
Methyl 6-bromo-1-(propan-2-yl)-1H-indazole-4-carboxylate
Methyl 6-bromo-1-(propan-2-yl)-1H-indazole-4-carboxylate
Formel:
C₁₂H₁₃BrN₂O₂ Molecular Weight: 297,148 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26407262 CAS-Nummer: 1346702-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...