5,5'-Dimethyl-2,2'-bipyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5,5'-Dimethyl-2,2'-bipyridine, Abametapir, 6,6'-Bi-3-picoline
OR31053-5G
OR31053-10G
OR31053-25G
APOSOR31053-5GEA
26.1
EUR
APOSOR31053-5G
APOSOR31053-10G
APOSOR31053-25G
5,5'-Dimethyl-2,2'-bipyridin
5,5'-Dimethyl-2,2'-bipyridyl
Formel:
C₁₂H₁₂N₂ Schmelzpunkt: 114…117 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01740554 CAS-Nummer: 1762-34-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...