Ethyl 5-(4-chlorophenyl)-1-methyl-1H-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR310530-1G
APOSOR310530-1GEA 0 EUR
APOSOR310530-1G
Ethyl 5-(4-chlorophenyl)-1-methyl-1H-pyrazole-3-carboxylate
Ethyl 5-(4-chlorophenyl)-1-methyl-1H-pyrazole-3-carboxylate
Formel: C₁₃H₁₃ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08446156
CAS-Nummer: 864426-88-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR