Ethyl 5-(3,5-dichlorophenyl)-1-methyl-1H-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR310536-1G
APOSOR310536-1GEA 538 EUR
APOSOR310536-1G
Ethyl 5-(3,5-dichlorophenyl)-1-methyl-1H-pyrazole-3-carboxylate
Ethyl 5-(3,5-dichlorophenyl)-1-methyl-1H-pyrazole-3-carboxylate
Formel: C₁₃H₁₂Cl₂N₂O₂
Molecular Weight: 299,153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27957340
CAS-Nummer: 1565827-84-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR