(1S,2R)-2-Phenylcyclopropan-1-amine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR310537-500MG
APOSOR310537-500MGEA 1350 EUR
APOSOR310537-500MG
(1S,2R)-2-Phenylcyclopropan-1-amine hydrochloride
(1S,2R)-2-Phenylcyclopropan-1-amine hydrochloride
Formel: C₉H₁₁N·ClH
Molecular Weight: 169,651 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21396228
CAS-Nummer: 4548-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR