(2R,3R)-3-[(2-Chlorophenyl)carbamoyl]-2,3-dihydroxypropanoic acid

Lieferant: Apollo Scientific

OR310538-1G
APOSOR310538-1GEA 894 EUR
APOSOR310538-1G
(2R,3R)-3-[(2-Chlorophenyl)carbamoyl]-2,3-dihydroxypropanoic acid
(2R,3R)-3-[(2-Chlorophenyl)carbamoyl]-2,3-dihydroxypropanoic acid
Formel: C₁₀H₁₀ClNO₅
Molecular Weight: 259,643 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27957341
CAS-Nummer: 17447-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR