(2S)-3-[(4-Methoxyphenyl)methoxy]-2-methylpropan-1-ol

Lieferant: Apollo Scientific
Achtung

OR310541-1G
APOSOR310541-1GEA 671 EUR
APOSOR310541-1G
(2S)-3-[(4-Methoxyphenyl)methoxy]-2-methylpropan-1-ol
(2S)-3-[(4-Methoxyphenyl)methoxy]-2-methylpropan-1-ol
Formel: C₁₂H₁₈O₃
Molecular Weight: 210,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27957342
CAS-Nummer: 160238-45-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR